COM448664272 Combi-Blocks, Inc.

6-Carboxyfluorescein hydrate

  • MDL number MFCD20257228
  • Cas Number : 3301-79-9
  • Molecular Weight : 394.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4744

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4744
Availability details for
Quantity
  • Availability
  • Add to Cart