COM448664291 Combi-Blocks, Inc.

1,3-Dichloro-5-methyl-2-nitrobenzene

  • MDL number MFCD22648298
  • Cas Number : 89692-81-9
  • Molecular Weight : 206.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4823

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4823
Availability details for
Quantity
  • Availability
  • Add to Cart