COM448664323 Combi-Blocks, Inc.

Bis-propargyl-peg6

  • MDL number MFCD26793755
  • Cas Number : 185378-83-0
  • Molecular Weight : 314.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4934

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4934
Availability details for
Quantity
  • Availability
  • Add to Cart