COM448664347 Combi-Blocks, Inc.

Bis-propargyl-PEG4

  • MDL number MFCD22683298
  • Cas Number : 126422-58-0
  • Molecular Weight : 226.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5053

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5053
Availability details for
Quantity
  • Availability
  • Add to Cart