COM448664436 Combi-Blocks, Inc.

Ethyl 3-bromo-7-oxo-4,5,6,7-tetrahydro-1h-indole-2-carboxylate

  • MDL number MFCD22741908
  • Cas Number : 174504-72-4
  • Molecular Weight : 286.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5397

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5397
Availability details for
Quantity
  • Availability
  • Add to Cart