COM448664484 Combi-Blocks, Inc.

Sodium 5-chlorobenzo[d]thiazole-2-carboxylate

  • MDL number MFCD23703168
  • Cas Number : 3507-53-7
  • Molecular Weight : 235.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5577

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5577
Availability details for
Quantity
  • Availability
  • Add to Cart