COM448664557 Combi-Blocks, Inc.

7-Quinolinesulfonyl chloride

  • MDL number MFCD19200224
  • Cas Number : 443692-93-1
  • Molecular Weight : 227.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5833

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5833
Availability details for
Quantity
  • Availability
  • Add to Cart