COM448664579 Combi-Blocks, Inc.

6,8-Dioxabicyclo[3.2.1]octan-3-one

  • MDL number MFCD22689934
  • Cas Number : 1369498-81-6
  • Molecular Weight : 128.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5923

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-5923
Availability details for
Quantity
  • Availability
  • Add to Cart