COM448664637 Combi-Blocks, Inc.

2,4-Difluoro-6-methoxybenzonitrile

  • MDL number MFCD20486220
  • Cas Number : 1240518-41-5
  • Molecular Weight : 169.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6138

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6138
Availability details for
Quantity
  • Availability
  • Add to Cart