COM448664643 Combi-Blocks, Inc.

3-Mercaptoisobutyric acid

  • MDL number MFCD00671555
  • Cas Number : 26473-47-2
  • Molecular Weight : 120.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6164

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6164
Availability details for
Quantity
  • Availability
  • Add to Cart