COM448664661 Combi-Blocks, Inc.

Cis-1,3-bis(aminomethyl)cyclohexane

  • MDL number MFCD19204670
  • Cas Number : 10340-00-8
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6232

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6232
Availability details for
Quantity
  • Availability
  • Add to Cart