COM448664676 Combi-Blocks, Inc.

7-Methoxy-6-nitro-1,2,3,4-tetrahydroisoquinoline HCl

  • MDL number MFCD27991309
  • Cas Number : 1820687-16-8
  • Molecular Weight : 244.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6311

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6311
Availability details for
Quantity
  • Availability
  • Add to Cart