COM448664677 Combi-Blocks, Inc.

6'-Bromospiro[cyclobutane-1,3'-indolin]-2'-one

  • MDL number MFCD19703308
  • Cas Number : 1190861-39-2
  • Molecular Weight : 252.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6313

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6313
Availability details for
Quantity
  • Availability
  • Add to Cart