COM448664700 Combi-Blocks, Inc.

8-Iodo-1h-benzo[d][1,3]oxazin-2(4h)-one

  • MDL number MFCD23703405
  • Cas Number : 1820612-81-4
  • Molecular Weight : 275.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6402

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6402
Availability details for
Quantity
  • Availability
  • Add to Cart