COM448664744 Combi-Blocks, Inc.

5-Bromo-3-isopropyl-2-methyl-3h-imidazo[4,5-b]pyridine

  • MDL number MFCD27991312
  • Cas Number : 1784871-38-0
  • Molecular Weight : 254.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6557

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6557
Availability details for
Quantity
  • Availability
  • Add to Cart