COM448664812 Combi-Blocks, Inc.

Methyl 2-amino-3-(1h-indol-4-yl)propanoate HCl

  • MDL number MFCD26406714
  • Cas Number : 1464091-39-1
  • Molecular Weight : 254.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6892

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6892
Availability details for
Quantity
  • Availability
  • Add to Cart