COM448664845 Combi-Blocks, Inc.

6-(Trifluoromethyl)-2h-spiro[benzofuran-3,4'-piperidine]

  • MDL number MFCD22581603
  • Cas Number : 721958-57-2
  • Molecular Weight : 257.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7031

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7031
Availability details for
Quantity
  • Availability
  • Add to Cart