COM448664896 Combi-Blocks, Inc.

Bromo-PEG5-t-butyl ester

  • MDL number MFCD22683304
  • Cas Number : 1415800-38-2
  • Molecular Weight : 429.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7222

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7222
Availability details for
Quantity
  • Availability
  • Add to Cart