COM448664913 Combi-Blocks, Inc.

Bis-propargyl-peg9

  • MDL number MFCD27635156
  • Cas Number : 1092554-87-4
  • Molecular Weight : 446.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7280

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7280
Availability details for
Quantity
  • Availability
  • Add to Cart