COM448665054 Combi-Blocks, Inc.

(Cis)-1-benzyl 2-methyl 4-oxocyclopentane-1,2-dicarboxylate

  • MDL number MFCD25542351
  • Cas Number : 164916-54-5
  • Molecular Weight : 276.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7853

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7853
Availability details for
Quantity
  • Availability
  • Add to Cart