COM448665095 Combi-Blocks, Inc.

Pramiracetam hydrate

  • MDL number MFCD27977197
  • Cas Number : 68497-62-1
  • Molecular Weight : 287.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8020

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8020
Availability details for
Quantity
  • Availability
  • Add to Cart