COM448665151 Combi-Blocks, Inc.

N,1-Dimethyl-1h-pyrazol-4-amine dihydrochloride

  • MDL number MFCD19689724
  • Cas Number : 948572-94-9
  • Molecular Weight : 184.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8218

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8218
Availability details for
Quantity
  • Availability
  • Add to Cart