COM448665155 Combi-Blocks, Inc.

(4-Iodo-6-methoxypyridin-3-yl)methanol

  • MDL number MFCD26403343
  • Cas Number : 1266728-78-2
  • Molecular Weight : 265.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8228

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-8228
Availability details for
Quantity
  • Availability
  • Add to Cart