COM448665360 Combi-Blocks, Inc.

4-Chloropiperidine

  • MDL number MFCD09997828
  • Cas Number : 5382-18-3
  • Molecular Weight : 119.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9019

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9019
Availability details for
Quantity
  • Availability
  • Add to Cart