COM448665396 Combi-Blocks, Inc.

1-Allylhydantoin

  • MDL number MFCD00142580
  • Cas Number : 3366-93-6
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9164

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9164
Availability details for
Quantity
  • Availability
  • Add to Cart