COM448665418 Combi-Blocks, Inc.

1-Bromo-4-methoxyisoquinoline

  • MDL number MFCD26406842
  • Cas Number : 1421517-83-0
  • Molecular Weight : 238.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9277

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9277
Availability details for
Quantity
  • Availability
  • Add to Cart