COM448665490 Combi-Blocks, Inc.

5-Carboxyfluorescein hydrate

  • MDL number MFCD20257226
  • Cas Number : 76823-03-5
  • Molecular Weight : 394.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9554

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-9554
Availability details for
Quantity
  • Availability
  • Add to Cart