COM448665611 Combi-Blocks, Inc.

Cetyltrimethylammonium hexafluorophosphate

  • MDL number MFCD00144894
  • Cas Number : 101079-29-2
  • Molecular Weight : 429.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-0111

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-0111
Availability details for
Quantity
  • Availability
  • Add to Cart