COM448665663 Combi-Blocks, Inc.

Cyclohexanecarboxylic acid isopropyl ester

  • MDL number MFCD00058949
  • Cas Number : 6553-80-6
  • Molecular Weight : 170.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-0884

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-0884
Availability details for
Quantity
  • Availability
  • Add to Cart