COM448665697 Combi-Blocks, Inc.

N-Bromosaccharin

  • MDL number MFCD03844772
  • Cas Number : 35812-01-2
  • Molecular Weight : 262.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-1287

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-1287
Availability details for
Quantity
  • Availability
  • Add to Cart