COM448665781 Combi-Blocks, Inc.

N-(2-Hydroxynaphthoyl)-2,4-dimethoxy-5-chloroanilide

  • MDL number MFCD00021635
  • Cas Number : 92-72-8
  • Molecular Weight : 357.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-2315

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-2315
Availability details for
Quantity
  • Availability
  • Add to Cart