COM448665885 Combi-Blocks, Inc.

4-tert-Butylthiacalix[4]arene

  • MDL number MFCD04038412
  • Cas Number : 182496-55-5
  • Molecular Weight : 721.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-3476

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-3476
Availability details for
Quantity
  • Availability
  • Add to Cart