COM448665923 Combi-Blocks, Inc.

2,2-Dimethyl-1,3-benzodioxole

  • MDL number MFCD08061594
  • Cas Number : 14005-14-2
  • Molecular Weight : 150.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-3939

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-3939
Availability details for
Quantity
  • Availability
  • Add to Cart