COM448665983 Combi-Blocks, Inc.

N-(3-Hydroxypropyl)ethylenediamine

  • MDL number MFCD00059848
  • Cas Number : 56344-32-2
  • Molecular Weight : 118.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-4618

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-4618
Availability details for
Quantity
  • Availability
  • Add to Cart