COM448666014 Combi-Blocks, Inc.

Diisopropylamine HCl

  • MDL number MFCD00034860
  • Cas Number : 819-79-4
  • Molecular Weight : 137.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-5021

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-5021
Availability details for
Quantity
  • Availability
  • Add to Cart