COM448666046 Combi-Blocks, Inc.

Succinaldehyde bis(dimethyl acetal)

  • MDL number MFCD00127971
  • Cas Number : 6922-39-0
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-5367

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-5367
Availability details for
Quantity
  • Availability
  • Add to Cart