COM448666053 Combi-Blocks, Inc.

Decanedioic acid disodium salt

  • MDL number MFCD00070501
  • Cas Number : 17265-14-4
  • Molecular Weight : 246.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-5468

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-5468
Availability details for
Quantity
  • Availability
  • Add to Cart