COM448666158 Combi-Blocks, Inc.

(2-Chlorobenzyl)triphenylphosphonium chloride

  • MDL number MFCD00054629
  • Cas Number : 18583-55-6
  • Molecular Weight : 423.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-6742

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-6742
Availability details for
Quantity
  • Availability
  • Add to Cart