COM448666176 Combi-Blocks, Inc.

Azelaic dihydrazide

  • MDL number MFCD00051529
  • Cas Number : 4080-95-9
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-6933

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-6933
Availability details for
Quantity
  • Availability
  • Add to Cart