COM448666197 Combi-Blocks, Inc.

Dodecylsuccinic anhydride

  • MDL number MFCD00022610
  • Cas Number : 2561-85-5
  • Molecular Weight : 268.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-7077

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-7077
Availability details for
Quantity
  • Availability
  • Add to Cart