COM448666328 Combi-Blocks, Inc.

3-Propoxypropylamine

  • MDL number MFCD00129001
  • Cas Number : 16728-59-9
  • Molecular Weight : 117.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-8716

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-8716
Availability details for
Quantity
  • Availability
  • Add to Cart