COM448666347 Combi-Blocks, Inc.

Sulfadimethoxine sodium salt

  • MDL number MFCD00070155
  • Cas Number : 1037-50-9
  • Molecular Weight : 332.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-9009

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-9009
Availability details for
Quantity
  • Availability
  • Add to Cart