COM448666356 Combi-Blocks, Inc.

3-(2-Hydroxyethoxy)benzaldehyde

  • MDL number MFCD00191448
  • Cas Number : 60345-97-3
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-9091

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-9091
Availability details for
Quantity
  • Availability
  • Add to Cart