COM448666364 Combi-Blocks, Inc.

Phenolphthalin

  • MDL number MFCD00036135
  • Cas Number : 81-90-3
  • Molecular Weight : 320.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-9208

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-9208
Availability details for
Quantity
  • Availability
  • Add to Cart