COM448666417 Combi-Blocks, Inc.

2,3-Dibromopropionamide

  • MDL number MFCD00031484
  • Cas Number : 15102-42-8
  • Molecular Weight : 230.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QE-9772

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QE-9772
Availability details for
Quantity
  • Availability
  • Add to Cart