COM448666558 Combi-Blocks, Inc.

1,2,5-Pentanetriol

  • MDL number MFCD01709258
  • Cas Number : 14697-46-2
  • Molecular Weight : 120.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-1428

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-1428
Availability details for
Quantity
  • Availability
  • Add to Cart