COM448666607 Combi-Blocks, Inc.

1-Sec-butyl-4-nitrobenzene

  • MDL number MFCD00130025
  • Cas Number : 4237-40-5
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2153

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2153
Availability details for
Quantity
  • Availability
  • Add to Cart