COM448666661 Combi-Blocks, Inc.

10-Amino-1-decanol

  • MDL number MFCD02094313
  • Cas Number : 23160-46-5
  • Molecular Weight : 173.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2740

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-2740
Availability details for
Quantity
  • Availability
  • Add to Cart