COM448666679 Combi-Blocks, Inc.

4-Ethyl-m-xylene

  • MDL number MFCD00039905
  • Cas Number : 874-41-9
  • Molecular Weight : 134.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3003

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3003
Availability details for
Quantity
  • Availability
  • Add to Cart