COM448666696 Combi-Blocks, Inc.

Cis-5-decene

  • MDL number MFCD00067488
  • Cas Number : 7433-78-5
  • Molecular Weight : 140.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3198

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3198
Availability details for
Quantity
  • Availability
  • Add to Cart